siRNA Details
Gene Symbol: HOXB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018952.4 | NP_061825.2 | 4 | 1517 | ||
XM_005257283.3 | XP_005257340.1 | 3 | 1759 | |||
XM_005257284.3 | XP_005257341.1 | 3 | 1815 | |||
XM_011524727.2 | XP_011523029.1 | 4 | 2013 | |||
GenBank mRNA | AI352188.1 | - | 1 | 41 | ||
AK292200.1 | - | 3 | 1815 | |||
BC014651.1 | - | 4 | 1517 | 4548382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
