Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC35C2 SIRNA Assays ›
Gene Symbol:
SLC35C2
Gene Name:
solute carrier family 35 member C2
Gene Aliases:
BA394O2.1, C20orf5, CGI-15, OVCOV1
Chromosome Location:
Chr.20: 46349528 - 46364461 on Build GRCh38
UniGene ID:
Hs.593344
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
23591
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC35C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281457.1
NP_001268386.1
8, 9
1119
NM_001281458.1
NP_001268387.1
9, 10
1058
NM_001281459.1
NP_001268388.1
8, 9
1056
NM_001281460.1
NP_001268389.1
9, 10
1522
NM_015945.11
NP_057029.8
8, 9
1190
NM_173073.3
NP_775096.1
8, 9
1127
NM_173179.3
NP_775271.1
8, 9
1385
XM_011528831.1
XP_011527133.1
8, 9
1368
XM_011528832.1
XP_011527134.1
8, 9
1662
XM_011528833.1
XP_011527135.1
8, 9
1186
XM_011528834.1
XP_011527136.1
8, 9
1187
XM_011528835.1
XP_011527137.1
7, 8
1076
XM_011528836.1
XP_011527138.1
8, 9
1271
XM_011528837.1
XP_011527139.1
8, 9
1105
XM_011528838.1
XP_011527140.1
6, 7
1224
XM_017027856.1
XP_016883345.1
8, 9
2085
XM_017027857.1
XP_016883346.1
8, 9
1613
XM_017027858.1
XP_016883347.1
8, 9
1523
XM_017027859.1
XP_016883348.1
8, 9
1890
XM_017027860.1
XP_016883349.1
9, 10
1330
XM_017027861.1
XP_016883350.1
8, 9
1289
XM_017027862.1
XP_016883351.1
8, 9
1228
GenBank mRNA
AF132949.1
-
8, 9
1127
AF455052.1
-
8, 9
1145
AK094358.1
-
1, 2
1152
AK128842.1
-
1, 2
2158
AK222928.1
-
8, 9
1130
AK300207.1
-
8, 9
1073
AK300798.1
-
8, 9
988
AK301022.1
-
9, 10
1000
AK315726.1
-
8, 9
1352
BC014191.2
-
8, 9
1071
4761663
BC021138.1
-
8, 9
1328
4870715
BC025277.1
-
8, 9
1386
4865888
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
negative regulation of gene expression
UDP-glucose transport
protein O-linked fucosylation
positive regulation of Notch signaling pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline