Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271848.1 | NP_001258777.1 | 4 | 1976 | ||
NM_144566.2 | NP_653167.1 | 4 | 1967 | |||
GenBank mRNA | AK126871.1 | - | 3 | 2188 | ||
AL136732.1 | - | 4 | 1950 | |||
BC136777.1 | - | 4 | 1894 | 9020767 | ||
BC144406.1 | - | 5 | 2015 |