Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSTYK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015375.2 | NP_056190.1 | 13 | 4077 | ||
NM_199462.2 | NP_955749.1 | 12 | 3942 | |||
XM_011509392.2 | XP_011507694.1 | 13 | 4008 | |||
XM_011509393.2 | XP_011507695.1 | 14 | 3971 | |||
XM_011509394.2 | XP_011507696.1 | 12 | 3479 | |||
GenBank mRNA | AB007941.1 | - | 8 | 2322 | ||
AY429674.1 | - | 13 | 4049 | |||
BC060870.1 | - | 7 | 3052 | 30346187 |