Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004948.3 | NP_004939.1 | 3 | 487 | ||
NM_024421.2 | NP_077739.1 | 3 | 487 | |||
GenBank mRNA | X72925.1 | - | 3 | 487 | ||
Z34522.1 | - | 3 | 415 |