siRNA Details
Gene Symbol: DNAJB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005494.2 | NP_005485.1 | 7 | 735 | ||
NM_058246.3 | NP_490647.1 | 7 | 734 | |||
XM_005249515.2 | XP_005249572.1 | 7 | 726 | |||
XM_005249516.2 | XP_005249573.1 | 7 | 870 | |||
XM_006715823.1 | XP_006715886.1 | 7 | 726 | |||
XM_011515704.1 | XP_011514006.1 | 7 | 705 | |||
GenBank mRNA | AB014888.1 | - | 7 | 680 | ||
AB015798.1 | - | 7 | 727 | |||
AB015799.1 | - | 7 | 727 | |||
AB209859.1 | - | 7 | 728 | |||
AF060703.1 | - | 7 | 725 | |||
AF075601.1 | - | 7 | 712 | |||
AK124827.1 | - | 2 | 1757 | |||
AK223601.1 | - | 7 | 745 | |||
AK291953.1 | - | 7 | 717 | |||
AK293025.1 | - | 7 | 718 | |||
AK297796.1 | - | 6 | 625 | |||
AK297989.1 | - | 7 | 594 | |||
AK301152.1 | - | 5 | 573 | |||
AL136707.1 | - | 7 | 739 | |||
AL832124.1 | - | 2 | 4351 | |||
BC000177.2 | - | 7 | 726 | 2900421 | ||
BC002446.2 | - | 7 | 734 | 3346070 | ||
BC043270.1 | - | 2 | 2446 | |||
CR533498.1 | - | 6 | 580 | |||
EU075177.1 | - | 7 | 724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
