siRNA Details
Gene Symbol: DNAJB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058246.3 | NP_490647.1 | 10 | 1104 | ||
XM_006715823.1 | XP_006715886.1 | 9 | 889 | |||
XM_017011633.1 | XP_016867122.1 | 7 | 772 | |||
GenBank mRNA | AB015798.1 | - | 10 | 1097 | ||
AB209859.1 | - | 11 | 6668 | |||
AK055429.1 | - | 1 | 1532 | |||
AK093837.1 | - | 1 | 734 | |||
AK223601.1 | - | 10 | 1115 | |||
AK293025.1 | - | 10 | 1088 | |||
AK297497.1 | - | 7 | 751 | |||
AK297796.1 | - | 9 | 995 | |||
AK297989.1 | - | 10 | 964 | |||
AK301152.1 | - | 8 | 943 | |||
BC002446.2 | - | 10 | 1104 | 3346070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
