Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018396.2 | NP_060866.2 | 9 | 1538 | ||
GenBank mRNA | AK002212.1 | - | 8 | 1412 | ||
AK299960.1 | - | 9 | 1502 | |||
BC064929.1 | - | 9 | 1498 | |||
BC107586.1 | - | 8 | 1422 | 6494734 |