Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018396.2 | NP_060866.2 | 9 | 1691 | ||
GenBank mRNA | AK002212.1 | - | 8 | 1566 | ||
BC064929.1 | - | 9 | 1652 | |||
BC107586.1 | - | 8 | 1576 | 6494734 |
Gene Symbol: METTL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181725.3 | NP_859076.3 | 9 | 1672 | ||
XM_017024572.1 | XP_016880061.1 | 8 | 1553 | |||
GenBank mRNA | AK000991.1 | - | 4 | 919 | ||
AK022822.1 | - | 9 | 1649 |