Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292018.1 | NP_001278947.1 | 14 | 1836 | ||
NM_001292019.1 | NP_001278948.1 | 14 | 1402 | |||
NM_004762.3 | NP_004753.1 | 14 | 1439 | |||
NM_017456.3 | NP_059430.2 | 13 | 1436 | |||
XM_011525475.2 | XP_011523777.1 | 13 | 2179 | |||
XM_011525476.2 | XP_011523778.1 | 14 | 1354 | |||
XM_011525477.1 | XP_011523779.1 | 12 | 1217 | |||
XM_011525478.1 | XP_011523780.1 | 13 | 1758 | |||
XM_017025336.1 | XP_016880825.1 | 13 | 1506 | |||
XM_017025337.1 | XP_016880826.1 | 14 | 1447 | |||
XM_017025338.1 | XP_016880827.1 | 13 | 1822 | |||
GenBank mRNA | AK091369.1 | - | 1 | 938 | ||
AK123894.1 | - | 1 | 2504 | |||
AK293894.1 | - | 14 | 1836 | |||
AK316075.1 | - | 14 | 1395 | |||
AK316277.1 | - | 14 | 1383 | |||
AL832791.1 | - | 1 | 1617 | |||
BC038385.1 | - | 14 | 1360 | 4755203 | ||
BC050452.1 | - | 14 | 1349 | 6043039 | ||
M85169.1 | - | 14 | 1389 |