siRNA Details
Gene Symbol: ATRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207047.2 | NP_001193976.1 | 22, 23 | 3269 | ||
NM_001323332.1 | NP_001310261.1 | 22, 23 | 3652 | |||
NM_139321.2 | NP_647537.1 | 22, 23 | 3652 | |||
NM_139322.3 | NP_647538.1 | 22, 23 | 3652 | |||
GenBank mRNA | AB011120.1 | - | 6, 7 | 633 | ||
AF106861.1 | - | 22, 23 | 3562 | |||
AK000356.1 | - | 7, 8 | 969 | |||
AK293010.1 | - | 22, 23 | 3652 | |||
AK302730.1 | - | 22, 23 | 3248 | |||
BC101705.1 | - | 22, 23 | 3602 | 8069211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
