Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301076.1 | NP_001288005.1 | 5 | 849 | ||
NM_001301078.1 | NP_001288007.1 | 5 | 966 | |||
NM_001301081.1 | NP_001288010.1 | 5 | 806 | |||
NM_004069.4 | NP_004060.2 | 5 | 924 | |||
NM_021575.3 | NP_067586.1 | 4 | 810 | |||
XM_011526423.2 | XP_011524725.1 | 5 | 902 | |||
XM_011526424.2 | XP_011524726.1 | 5 | 797 | |||
GenBank mRNA | BC006337.2 | - | 5 | 733 | 4304127 | |
BU564736.1 | - | 140 | ||||
X97074.1 | - | 5 | 811 |