siRNA Details
Gene Symbol: SOX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308165.1 | NP_001295094.1 | 6 | 1553 | ||
NM_007017.2 | NP_008948.1 | 4 | 2163 | |||
NM_178424.1 | NP_848511.1 | 5 | 2656 | |||
XM_005265803.4 | XP_005265860.1 | 6 | 1420 | |||
XM_011534420.1 | XP_011532722.1 | 6 | 2446 | |||
GenBank mRNA | AB022083.1 | - | 5 | 2609 | ||
AB022441.1 | - | 4 | 2116 | |||
AK302162.1 | - | 6 | 1553 | |||
AK308329.1 | - | 5 | 2604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
