Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006710814.3 | XP_006710877.1 | 9, 10 | 2840 | ||
XM_017002050.1 | XP_016857539.1 | 9, 10 | 2690 | |||
GenBank mRNA | AF294278.1 | - | 9, 10 | 2671 |