siRNA Details
Gene Symbol: NOTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017617.4 | NP_060087.3 | 34 | 8960 | ||
GenBank mRNA | AB209873.1 | - | 26 | 7497 | ||
AK000012.1 | - | 1 | 1136 | |||
BC013208.1 | - | 4 | 3145 | |||
BC046127.1 | - | 1 | 711 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -




