Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004501.3 | NP_004492.2 | 14 | 3462 | ||
NM_031844.2 | NP_114032.2 | 14 | 3519 | |||
XM_017001115.1 | XP_016856604.1 | 14 | 3714 | |||
XM_017001116.1 | XP_016856605.1 | 14 | 3519 | |||
XM_017001117.1 | XP_016856606.1 | 14 | 3508 | |||
GenBank mRNA | AK095525.1 | - | 1 | 45 | ||
AK126868.1 | - | 15 | 3260 | |||
BC015782.1 | - | 14 | 2996 | 4857112 |