Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146077.1 | NP_001139549.1 | 3 | 1304 | ||
NM_001146078.2 | NP_001139550.1 | 3 | 1400 | |||
NM_001146079.1 | NP_001139551.1 | 2 | 1603 | |||
NM_012130.3 | NP_036262.1 | 2 | 1082 | |||
NM_144492.2 | NP_652763.1 | 3 | 1793 | |||
GenBank mRNA | AF314090.1 | - | 3 | 1793 | ||
AJ566765.1 | - | 3 | 1181 | |||
AJ566766.1 | - | 3 | 1304 | |||
AY355348.1 | - | 2 | 1603 | |||
AY355349.1 | - | 2 | 1082 | |||
AY358533.1 | - | 1 | 1015 | |||
BC012126.1 | - | 2 | 1068 | 4684949 |