Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LARP4 SIRNA Assays ›
Gene Symbol:
LARP4
Gene Name:
La ribonucleoprotein domain family member 4
Gene Aliases:
PP13296
Chromosome Location:
Chr.12: 50400809 - 50480005 on Build GRCh38
UniGene ID:
Hs.26613
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
236804
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LARP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170804.1
NP_001164275.1
11
1401
NM_052879.4
NP_443111.4
11
1401
NM_199188.2
NP_954658.2
11
1398
NM_199190.2
NP_954660.1
10
1188
NR_033200.1
12
1450
XM_005268613.2
XP_005268670.1
12
1450
XM_011537828.2
XP_011536130.1
12
1505
XM_011537829.2
XP_011536131.1
12
1486
XM_011537830.2
XP_011536132.1
11
1333
XM_011537831.2
XP_011536133.1
11
1397
XM_011537833.2
XP_011536135.1
11
1292
XM_011537835.2
XP_011536137.1
11
1273
XM_011537838.2
XP_011536140.1
11
1245
XM_011537839.2
XP_011536141.1
12
1389
XM_011537840.2
XP_011536142.1
12
1442
XM_011537841.2
XP_011536143.1
11
1295
XM_011537844.2
XP_011536146.1
12
1506
XM_017018750.1
XP_016874239.1
10
1114
XM_017018751.1
XP_016874240.1
12
1266
XM_017018752.1
XP_016874241.1
11
1249
XM_017018753.1
XP_016874242.1
11
1282
XM_017018754.1
XP_016874243.1
12
1432
XM_017018755.1
XP_016874244.1
10
1099
XM_017018756.1
XP_016874245.1
11
1151
XM_017018757.1
XP_016874246.1
12
1321
XM_017018758.1
XP_016874247.1
11
1257
XM_017018760.1
XP_016874249.1
11
1401
XM_017018761.1
XP_016874250.1
10
1181
GenBank mRNA
AF370416.1
-
11
1279
AK055521.1
-
9
949
AK125113.1
-
10
1191
AK291746.1
-
11
1270
AL833272.1
-
11
1311
AY004310.1
-
12
1305
BC063479.1
-
11
1301
5219803
BX647457.1
-
10
1012
CR936626.1
-
11
1319
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
regulation of cell morphogenesis
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0