Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVSSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317934.1 | NP_001304863.1 | 11 | 2018 | ||
NM_001317935.1 | NP_001304864.1 | 11 | 2006 | |||
NM_020894.3 | NP_065945.2 | 11 | 2119 | |||
XM_017008490.1 | XP_016863979.1 | 11 | 1840 | |||
XM_017008491.1 | XP_016863980.1 | 11 | 1932 | |||
XM_017008492.1 | XP_016863981.1 | 11 | 1930 | |||
XM_017008493.1 | XP_016863982.1 | 11 | 1806 | |||
XM_017008494.1 | XP_016863983.1 | 11 | 1691 | |||
XM_017008495.1 | XP_016863984.1 | 8 | 1833 | |||
XM_017008500.1 | XP_016863989.1 | 5 | 560 | |||
GenBank mRNA | AB040963.1 | - | 11 | 2119 | ||
AK292661.1 | - | 4 | 739 | |||
AL390128.1 | - | 2 | 239 | |||
BC021930.1 | - | 4 | 757 | |||
BC110331.1 | - | 11 | 2018 | 5739956 | ||
BC140901.1 | - | 11 | 1856 | 9021769 | ||
EL735271.1 | - | 3 | 325 |