Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300728.1 | NP_001287657.1 | 4 | 2331 | ||
NM_015251.2 | NP_056066.2 | 4 | 2391 | |||
GenBank mRNA | AB007891.1 | - | 3 | 2868 | ||
AK025058.1 | - | 1 | 229 | |||
AK290943.1 | - | 4 | 2176 | |||
AK301781.1 | - | 3 | 2131 | |||
BC002701.2 | - | 4 | 2208 | |||
BM693775.1 | - | 1 | 285 | |||
CR749457.1 | - | 4 | 2284 |