Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIDT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040455.1 | NP_001035545.1 | 13 | 1270 | ||
GenBank mRNA | AF151799.1 | - | 14 | 1375 | ||
AK075137.1 | - | 4 | 248 | |||
AK292501.1 | - | 13 | 1750 | |||
AK292974.1 | - | 13 | 1261 | |||
AK301501.1 | - | 5 | 540 | |||
AK303023.1 | - | 13 | 1226 | |||
AY358442.1 | - | 13 | 1279 | |||
BC114522.1 | - | 14 | 1290 | 40035836 | ||
BX648437.1 | - | 5 | 368 | |||
KC109742.1 | - | 13 | 1226 |