siRNA Details
Gene Symbol: SNCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001502.2 | NP_001001502.1 | 7 | 1252 | ||
NM_001318034.1 | NP_001304963.1 | 6 | 1210 | |||
NM_001318035.1 | NP_001304964.1 | 6 | 1162 | |||
NM_001318036.1 | NP_001304965.1 | 5 | 1053 | |||
NM_001318037.1 | NP_001304966.1 | 5 | 1005 | |||
NM_003085.4 | NP_003076.1 | 6 | 1095 | |||
XM_006714916.2 | XP_006714979.1 | 6 | 1060 | |||
GenBank mRNA | AB209029.1 | - | 3 | 2645 | ||
BC002902.2 | - | 7 | 1055 | 3944215 | ||
JF960950.1 | - | 6 | 1205 | |||
JF960951.1 | - | 6 | 1157 | |||
JF960952.1 | - | 5 | 1048 | |||
JF960953.1 | - | 5 | 1000 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
