siRNA Details
Gene Symbol: SNCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001502.2 | NP_001001502.1 | 6, 7 | 831 | ||
NM_001318034.1 | NP_001304963.1 | 5, 6 | 789 | |||
NM_001318035.1 | NP_001304964.1 | 5, 6 | 741 | |||
NM_001318036.1 | NP_001304965.1 | 4, 5 | 632 | |||
NM_001318037.1 | NP_001304966.1 | 4, 5 | 584 | |||
NM_003085.4 | NP_003076.1 | 5, 6 | 674 | |||
XM_006714916.2 | XP_006714979.1 | 5, 6 | 639 | |||
GenBank mRNA | BC002902.2 | - | 6, 7 | 634 | 3944215 | |
BM543516.1 | - | 5, 6 | 674 | |||
BM967045.1 | - | 2, 3 | 202 | |||
BT006627.1 | - | 4, 5 | 383 | |||
CR457027.1 | - | 4, 5 | 383 | |||
JF960950.1 | - | 5, 6 | 784 | |||
JF960951.1 | - | 5, 6 | 736 | |||
JF960952.1 | - | 4, 5 | 627 | |||
JF960953.1 | - | 4, 5 | 579 | |||
S69965.1 | - | 5, 6 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
