Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321102.1 | NP_001308031.1 | 10 | 2028 | ||
NM_016030.5 | NP_057114.5 | 10 | 2065 | |||
XM_011510350.2 | XP_011508652.1 | 9 | 2381 | |||
XM_011510355.2 | XP_011508657.1 | 9 | 1006 | |||
XM_011510356.1 | XP_011508658.1 | 9 | 898 | |||
GenBank mRNA | AF151845.1 | - | 8 | 794 | ||
AK094181.1 | - | 11 | 2033 | |||
AK094242.1 | - | 9 | 1439 | |||
AK098327.1 | - | 10 | 2030 | |||
AK125105.1 | - | 5 | 2945 | |||
BC014164.2 | - | 10 | 1942 | 4548681 | ||
BC017475.1 | - | 9 | 1063 | |||
BQ954367.1 | - | 5 | 377 |