Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016128.3 | NP_057212.1 | 3 | 212 | ||
XM_011512549.2 | XP_011510851.1 | 3 | 247 | |||
GenBank mRNA | AB047846.1 | - | 3 | 127 | ||
AF100756.1 | - | 3 | 187 | |||
AK001724.1 | - | 3 | 190 | |||
AK291693.1 | - | 3 | 216 | |||
AK309821.1 | - | 3 | 190 | |||
BC066650.1 | - | 3 | 175 | 6046736 | ||
BU521557.1 | - | 213 |