Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286839.1 | NP_001273768.1 | 15 | 1868 | ||
NM_005766.3 | NP_005757.1 | 15 | 2047 | |||
XM_011521046.2 | XP_011519348.1 | 15 | 2047 | |||
XM_017020312.1 | XP_016875801.1 | 15 | 1791 | |||
XM_017020313.1 | XP_016875802.1 | 14 | 2784 | |||
GenBank mRNA | AB008430.1 | - | 15 | 1677 | ||
BC041595.1 | - | 15 | 1894 | 5443230 | ||
BC071592.1 | - | 15 | 1868 | 30342239 |