Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 32 | 6900 | ||
NM_001265611.1 | NP_001252540.1 | 30 | 6051 | |||
NM_014994.2 | NP_055809.2 | 31 | 6882 | |||
NR_049761.1 | 30 | 6692 | ||||
NR_049762.1 | 29 | 6643 | ||||
XM_006720438.1 | XP_006720501.1 | 30 | 6699 | |||
XM_006720439.2 | XP_006720502.1 | 18 | 4974 | |||
XM_011521383.1 | XP_011519685.1 | 31 | 6717 | |||
XM_017022017.1 | XP_016877506.1 | 30 | 6993 |