Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024940.2 | NP_001020111.1 | 3 | 798 | ||
NM_001134855.1 | NP_001128327.1 | 3 | 950 | |||
NM_016102.3 | NP_057186.1 | 3 | 834 | |||
XM_006711779.2 | XP_006711842.1 | 3 | 861 | |||
XM_011544209.2 | XP_011542511.1 | 3 | 848 | |||
XM_011544210.2 | XP_011542512.1 | 3 | 950 | |||
XM_011544211.2 | XP_011542513.1 | 3 | 851 | |||
XM_017001418.1 | XP_016856907.1 | 2 | 1406 | |||
XM_017001419.1 | XP_016856908.1 | 3 | 950 | |||
GenBank mRNA | AF156271.1 | - | 3 | 557 | ||
AK055426.1 | - | 2 | 1406 | |||
AK301105.1 | - | 3 | 754 | |||
AK308774.1 | - | 3 | 825 | |||
AK316379.1 | - | 3 | 585 | |||
BC033788.1 | - | 3 | 598 | 5165609 | ||
BI825739.1 | - | 3 | 582 | |||
BM563820.1 | - | 3 | 480 |