Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000757.5 | NP_000748.3 | 2 | 532 | ||
NM_172210.2 | NP_757349.1 | 2 | 532 | |||
NM_172211.3 | NP_757350.1 | 2 | 532 | |||
NM_172212.2 | NP_757351.1 | 2 | 532 | |||
XM_017000369.1 | XP_016855858.1 | 2 | 207 | |||
GenBank mRNA | AB208952.1 | - | 2 | 281 | ||
AK291660.1 | - | 2 | 294 | |||
BC021117.1 | - | 2 | 308 | 4586312 | ||
DA414355.1 | - | 2 | 532 | |||
DB287787.1 | - | 2 | 465 | |||
JF803735.1 | - | 1 | 23 | |||
M27087.1 | - | 2 | 243 | |||
M37435.1 | - | 1 | 224 | |||
M64592.1 | - | 2 | 119 | |||
M76453.1 | - | 1 | 83 | |||
U22386.1 | - | 2 | 163 | |||
X05825.1 | - | 1 | 67 |