siRNA Details
Gene Symbol: MT1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175617.3 | NP_783316.2 | 3 | 473 | ||
GenBank mRNA | AF495759.1 | - | 2 | 688 | ||
BC009699.2 | - | 3 | 671 | 3892441 | ||
BC062734.1 | - | 3 | 370 | 6560167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
