Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ING4 SIRNA Assays ›
          Gene Symbol:
          ING4
          Gene Name:
          inhibitor of growth family member 4
          Gene Aliases:
          my036, p29ING4
          Chromosome Location:
          Chr.12: 6650538 - 6663143 on Build GRCh38
          UniGene ID:
          Hs.524210
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 23834

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ING4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127582.1 NP_001121054.1 2 135
          NM_001127583.1 NP_001121055.1 2 135
          NM_001127584.1 NP_001121056.1 2 135
          NM_001127586.1 NP_001121058.1 2 135
          NM_016162.3 NP_057246.2 2 135
          XM_005253698.3 XP_005253755.1 2 107
          XM_011520965.2 XP_011519267.1 2 82
          XM_017019392.1 XP_016874881.1 2 107
          XM_017019393.1 XP_016874882.1 2 107
          GenBank mRNA AB197695.1 - 2 94
          AB197696.1 - 2 94
          AB197697.1 - 2 94
          AF063594.1 - 2 113
          AF110645.1 - 2 111
          AF156552.1 - 2 111
          AK303354.1 - 2 117
          AK313621.1 - 2 107
          BC007781.2 - 2 97 4309278
          BC013038.2 - 2 80
          BC095434.1 - 2 99 30365729
          BG705367.1 - 2 135
          BG718170.1 - 2 146
          BM922497.1 - 2 88
          BQ066994.1 - 106
          EF152350.1 - 2 94
          EF152351.1 - 2 94
          EF152352.1 - 2 94

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          protein acetylation
          apoptotic process
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          cell cycle arrest
          negative regulation of cell proliferation
          chromatin modification
          positive regulation of apoptotic process
          histone H3 acetylation
          histone H4-K5 acetylation
          histone H4-K8 acetylation
          histone H4-K12 acetylation
          histone H4-K16 acetylation
          negative regulation of transcription, DNA-templated
          negative regulation of growth
          transcription coactivator activity
          protein binding
          zinc ion binding
          methylated histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0