Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006910.4 | NP_008841.2 | 17 | 3441 | ||
NM_018703.3 | NP_061173.1 | 16 | 3339 | |||
GenBank mRNA | AB112074.1 | - | 17 | 2445 | ||
AB112075.1 | - | 16 | 2343 | |||
AF116625.1 | - | 2 | 679 | |||
AF352051.1 | - | 12 | 1964 | |||
AK303040.1 | - | 15 | 2339 | |||
AY072922.1 | - | 12 | 3088 | |||
BC015318.1 | - | 1 | 315 | |||
BC030964.1 | - | 2 | 645 | |||
BX648661.1 | - | 17 | 2381 | |||
X85133.1 | - | 12 | 1964 |