Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBOX1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125766.1 | 1 | 375 | |||
GenBank mRNA | DA376689.1 | - | 1 | 27 |
Gene Symbol: CASC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103848.1 | 1 | 76 | |||
NR_103850.2 | 1 | 76 | ||||
GenBank mRNA | CN418510.1 | - | 1 | 62 |
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143970.1 | NP_001137442.1 | 1 | 96 | ||
GenBank mRNA | AK296985.1 | - | 1 | 96 |
Gene Symbol: CHRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000740.2 | NP_000731.1 | 1 | 418 | ||
GenBank mRNA | AF279779.1 | - | 1 | 417 |
Gene Symbol: FOCAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017794.4 | NP_060264.4 | 2 | 227 | ||
XM_017014855.1 | XP_016870344.1 | 1 | 64 | |||
GenBank mRNA | AK056522.1 | - | 2 | 227 | ||
AY139834.1 | - | 2 | 226 |
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF279774.1 | - | 2 | 288 |
Gene Symbol: LINC00381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA421131.1 | - | 3 | 312 |
Gene Symbol: LINC00491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103753.1 | 1 | 199 | |||
NR_103754.1 | 1 | 199 | ||||
NR_103755.1 | 1 | 199 | ||||
NR_103756.1 | 1 | 199 | ||||
GenBank mRNA | BG777293.1 | - | 1 | 150 | ||
DA458115.1 | - | 1 | 18 | |||
DA458823.1 | - | 1 | 199 |
Gene Symbol: LOC105378763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF279783.1 | - | 2 | 300 |
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026981.1 | 1 | 200 | |||
GenBank mRNA | BQ668440.1 | - | 1 | 200 |
Gene Symbol: SLC44A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296924.1 | - | 1 | 139 | ||
AK296941.1 | - | 1 | 45 |
Gene Symbol: SNCAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543741.2 | XP_011542043.1 | 1 | 30 |