Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC107983983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022769.1 | XP_016878258.1 | 2 | 250 |
Gene Symbol: IGHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130817.1 | - | 2 | 165 |
Gene Symbol: LOC102724971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543804.2 | XP_011542106.1 | 2 | 244 |