Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007525.3 | NP_001007526.3 | 19 | 5717 | ||
NM_001290355.1 | NP_001277284.1 | 21 | 6475 | |||
XM_011527929.1 | XP_011526231.1 | 21 | 6007 | |||
XM_011527930.2 | XP_011526232.1 | 21 | 6493 | |||
XM_011527931.2 | XP_011526233.1 | 20 | 6273 | |||
XM_011527934.1 | XP_011526236.1 | 16 | 4212 | |||
XM_017026669.1 | XP_016882158.1 | 22 | 6560 | |||
GenBank mRNA | AK026798.1 | - | 1 | 233 | ||
AK090685.1 | - | 3 | 1441 | |||
BC117698.1 | - | 19 | 5471 | 40073616 | ||
BX648940.1 | - | 22 | 6475 | |||
CR749794.1 | - | 19 | 5713 |