siRNA Details
Gene Symbol: TEX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003811.1 | NP_001003811.1 | 30 | 2809 | ||
NM_031276.2 | NP_112566.2 | 29 | 2687 | |||
XM_011530994.1 | XP_011529296.1 | 29 | 2681 | |||
XM_011530996.1 | XP_011529298.1 | 18 | 1827 | |||
XM_017029649.1 | XP_016885138.1 | 29 | 2681 | |||
XM_017029650.1 | XP_016885139.1 | 18 | 1873 | |||
XM_017029651.1 | XP_016885140.1 | 18 | 1827 | |||
XM_017029652.1 | XP_016885141.1 | 16 | 1508 | |||
GenBank mRNA | AF285594.1 | - | 28 | 2592 | ||
AK057391.1 | - | 18 | 1828 | |||
AK057523.1 | - | 30 | 2803 | |||
AK292471.1 | - | 29 | 2681 | |||
BC036016.1 | - | 29 | 2687 | 5270970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
