Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC729218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103825.1 | 3 | 840 | |||
NR_109983.1 | 10 | 1783 | ||||
GenBank mRNA | AK024248.1 | - | 10 | 1754 |
Gene Symbol: LINC01061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037596.1 | 3 | 850 |