Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079870.1 | NP_001073339.1 | 5 | 1022 | ||
NM_001079871.1 | NP_001073340.1 | 5 | 998 | |||
NM_177977.2 | NP_817084.2 | 5 | 998 | |||
GenBank mRNA | AB209105.1 | - | 5 | 983 | ||
AF040723.1 | - | 5 | 996 | |||
AK022007.1 | - | 5 | 1019 | |||
U38371.1 | - | 2 | 224 |