siRNA Details
Gene Symbol: SIRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193286.1 | NP_001180215.1 | 5 | 614 | ||
NM_012237.3 | NP_036369.2 | 6 | 622 | |||
NM_030593.2 | NP_085096.1 | 5 | 614 | |||
NR_034146.1 | 4 | 545 | ||||
XM_006723111.1 | XP_006723174.1 | 7 | 570 | |||
XM_011526654.1 | XP_011524956.1 | 5 | 507 | |||
XM_011526655.1 | XP_011524957.1 | 3 | 322 | |||
XM_017026500.1 | XP_016881989.1 | 5 | 378 | |||
GenBank mRNA | AB009274.1 | - | 5 | 338 | ||
AF083107.2 | - | 6 | 520 | |||
AF095714.1 | - | 5 | 393 | |||
AF131800.1 | - | 4 | 269 | |||
AF160214.1 | - | 5 | 374 | |||
AJ505014.1 | - | 3 | 1091 | |||
AK025876.1 | - | 5 | 400 | |||
AK054642.1 | - | 6 | 453 | |||
AK131390.1 | - | 2 | 930 | |||
AK225824.1 | - | 5 | 397 | |||
AK290716.1 | - | 4 | 545 | |||
AK314492.1 | - | 6 | 376 | |||
AY030277.1 | - | 5 | 375 | |||
BC003012.1 | - | 5 | 465 | 2820929 | ||
BC003547.1 | - | 5 | 465 | 3536728 | ||
KF032391.1 | - | 3 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -





