siRNA Details
Gene Symbol: APOL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030643.4 | NP_085146.2 | 4 | 473 | ||
NM_145660.2 | NP_663693.1 | 3 | 284 | |||
GenBank mRNA | AF305226.1 | - | 2 | 125 | ||
AK225696.1 | - | 2 | 134 | |||
AK290466.1 | - | 3 | 458 | |||
AK290777.1 | - | 2 | 105 | |||
AK311506.1 | - | 3 | 257 | |||
AK312716.1 | - | 2 | 183 | |||
AK313637.1 | - | 3 | 221 | |||
AY014914.1 | - | 4 | 474 | |||
AY014915.1 | - | 3 | 284 | |||
BC006276.2 | - | 5 | 436 | |||
CR456380.1 | - | 2 | 63 | |||
DA865632.1 | - | 3 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
