Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF628 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033113.2 | NP_149104.3 | 3 | 782 | ||
XM_005259371.3 | XP_005259428.1 | 2 | 427 | |||
GenBank mRNA | BC089449.1 | - | 3 | 782 | 30560806 |