Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008410.1 | NP_001008410.1 | 4 | 1190 | ||
NM_018234.2 | NP_060704.2 | 5 | 1258 | |||
NM_138637.2 | NP_619543.2 | 4 | 1190 | |||
NM_182915.2 | NP_878919.2 | 5 | 1605 | |||
XM_006712614.3 | XP_006712677.1 | 5 | 1332 | |||
XM_006712615.1 | XP_006712678.1 | 4 | 1202 | |||
XM_011511403.1 | XP_011509705.1 | 6 | 1616 | |||
GenBank mRNA | AF238864.1 | - | 4 | 1146 | ||
AF262322.1 | - | 4 | 1146 | |||
AF423424.1 | - | 5 | 1235 | |||
AK001691.1 | - | 5 | 1235 | |||
AK024163.1 | - | 2 | 223 | |||
AK291608.1 | - | 5 | 1567 | |||
AL833624.1 | - | 5 | 1605 | |||
AY029585.1 | - | 5 | 1235 | |||
AY082673.1 | - | 4 | 1142 | |||
AY214461.1 | - | 5 | 1235 | |||
BC042150.1 | - | 4 | 1141 | 4586693 | ||
BC095421.1 | - | 4 | 1145 | 30336214 | ||
BX538047.1 | - | 5 | 1574 |