Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012133.5 | NP_036265.3 | 23 | 2542 | ||
GenBank mRNA | AB047847.1 | - | 23 | 2452 | ||
AF157833.1 | - | 23 | 2512 | |||
AF207598.1 | - | 23 | 2508 | |||
AF308295.1 | - | 4 | 405 | |||
AK002086.1 | - | 15 | 1844 | |||
AK022934.1 | - | 23 | 2439 | |||
AK024414.1 | - | 16 | 1920 | |||
AK291409.1 | - | 23 | 2532 | |||
BC017443.1 | - | 23 | 2435 |