Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301107.1 | NP_001288036.1 | 5 | 521 | ||
NM_016053.3 | NP_057137.1 | 5 | 524 | |||
XM_011538423.1 | XP_011536725.1 | 5 | 442 | |||
XM_017019383.1 | XP_016874872.1 | 4 | 339 | |||
GenBank mRNA | AF151874.1 | - | 5 | 469 | ||
AF155655.1 | - | 5 | 469 | |||
AK223339.1 | - | 5 | 460 | |||
AK314970.1 | - | 5 | 442 | |||
BC010889.1 | - | 6 | 395 | 4272385 | ||
BX356987.2 | - | 5 | 476 | |||
CR457171.1 | - | 5 | 362 |