Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318498.1 | NP_001305427.1 | 9 | 992 | ||
NM_001318499.1 | NP_001305428.1 | 9 | 1057 | |||
NM_001318500.1 | NP_001305429.1 | 6 | 661 | |||
NM_016055.5 | NP_057139.1 | 8 | 934 | |||
NR_134658.1 | 7 | 896 | ||||
NR_134659.1 | 10 | 1187 | ||||
NR_134660.1 | 8 | 1052 | ||||
NR_134661.1 | 6 | 742 | ||||
XM_017017919.1 | XP_016873408.1 | 9 | 1027 | |||
GenBank mRNA | AF151876.1 | - | 8 | 1490 | ||
AF161408.1 | - | 8 | 937 | |||
AK297746.1 | - | 9 | 992 | |||
AK310005.1 | - | 10 | 1166 | |||
BC009764.1 | - | 8 | 927 | 4093230 |