Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291920.1 | NP_001278849.1 | 10 | 5503 | ||
NM_001291921.1 | NP_001278850.1 | 9 | 5294 | |||
NM_002957.5 | NP_002948.1 | 10 | 4831 | |||
GenBank mRNA | AF052092.1 | - | 1 | 839 | ||
BC007925.1 | - | 1 | 1754 | |||
BC063827.1 | - | 10 | 4820 | |||
U66306.1 | - | 1 | 3082 |