siRNA Details
Gene Symbol: HIST1H3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003529.2 | NP_003520.1 | 1 | 408 | ||
GenBank mRNA | BC066245.1 | - | 1 | 408 | 7002068 | |
BC066246.1 | - | 1 | 408 | 7002069 | ||
BC066247.1 | - | 1 | 408 | 7002070 | ||
BC067490.1 | - | 1 | 408 | 7002071 | ||
BC067491.1 | - | 1 | 408 | 7002072 | ||
BC069303.1 | - | 1 | 408 | 7262697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
