siRNA Details
Gene Symbol: NUDT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301022.1 | NP_001287951.1 | 5 | 744 | ||
NM_001301023.1 | NP_001287952.1 | 5 | 741 | |||
NM_001301024.1 | NP_001287953.1 | 5 | 644 | |||
NM_019094.5 | NP_061967.3 | 5 | 999 | |||
NM_199040.3 | NP_950241.1 | 5 | 1002 | |||
XM_011537794.2 | XP_011536096.1 | 5 | 542 | |||
XM_011537795.2 | XP_011536097.1 | 6 | 2554 | |||
XM_011537796.2 | XP_011536098.1 | 6 | 683 | |||
GenBank mRNA | AB007956.2 | - | 6 | 798 | ||
AF067803.1 | - | 7 | 621 | |||
AF191649.1 | - | 5 | 772 | |||
AF191650.1 | - | 5 | 611 | |||
AF191651.1 | - | 5 | 590 | |||
AF191652.2 | - | 5 | 593 | |||
AF191653.1 | - | 1 | 775 | |||
AK223497.1 | - | 5 | 944 | |||
AK304296.1 | - | 5 | 716 | |||
BC012069.1 | - | 5 | 792 | 4650281 | ||
BC051310.1 | - | 5 | 678 | 5814319 | ||
CN335956.1 | - | 5 | 578 | |||
CR749445.1 | - | 5 | 714 |
Gene Symbol: NUDT4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002212.4 | 1 | 955 | |||
GenBank mRNA | AF311215.1 | - | LIT2089 NUDT4 pseudogene |
5 | 876 |
Gene Symbol: NUDT4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_104005.1 | 1 | 999 | |||
GenBank mRNA | AF311216.1 | - | LIT2090 NUDT4 pseudogene |
1 | 661 | |
AK074343.1 | - | 5 | 1001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
