Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301022.1 | NP_001287951.1 | 5 | 691 | ||
NM_001301023.1 | NP_001287952.1 | 5 | 688 | |||
NM_001301024.1 | NP_001287953.1 | 5 | 591 | |||
NM_019094.5 | NP_061967.3 | 5 | 946 | |||
NM_199040.3 | NP_950241.1 | 5 | 949 | |||
XM_011537794.2 | XP_011536096.1 | 5 | 489 | |||
XM_011537795.2 | XP_011536097.1 | 6 | 2501 | |||
XM_011537796.2 | XP_011536098.1 | 6 | 630 | |||
GenBank mRNA | AB007956.2 | - | 6 | 745 | ||
AF191649.1 | - | 5 | 719 | |||
AF191650.1 | - | 5 | 558 | |||
AF191651.1 | - | 5 | 537 | |||
AF191652.2 | - | 5 | 540 | |||
AF191653.1 | - | 1 | 722 | |||
AK223497.1 | - | 5 | 891 | |||
AK304296.1 | - | 5 | 663 | |||
BC012069.1 | - | 5 | 739 | 4650281 | ||
BC051310.1 | - | 5 | 625 | 5814319 | ||
BP315657.1 | - | 5 | 563 | |||
CR749445.1 | - | 5 | 661 |
Gene Symbol: NUDT4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002212.4 | 1 | 902 | |||
GenBank mRNA | AF311215.1 | - | LIT2089 NUDT4 pseudogene |
5 | 823 |
Gene Symbol: NUDT4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_104005.1 | 1 | 946 | |||
GenBank mRNA | AF311216.1 | - | LIT2090 NUDT4 pseudogene |
1 | 608 | |
AK074343.1 | - | 5 | 948 |