Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297438.1 | NP_001284367.1 | 3 | 630 | ||
NM_014231.4 | NP_055046.1 | 3 | 630 | |||
NM_016830.3 | NP_058439.1 | 3 | 630 | |||
NM_199245.2 | NP_954740.1 | 3 | 630 | |||
NR_123717.1 | 2 | 503 | ||||
GenBank mRNA | AF060538.1 | - | 3 | 392 | ||
AK289498.1 | - | 3 | 401 | |||
AK296294.1 | - | 3 | 408 | |||
AK303137.1 | - | 3 | 408 | |||
BI838752.1 | - | 3 | 425 | |||
BQ717992.1 | - | 3 | 389 | |||
CR542214.1 | - | 2 | 262 | |||
CR542231.1 | - | 2 | 262 | |||
CR749485.1 | - | 3 | 631 | |||
CR989458.1 | - | 3 | 384 |